Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043281.2 | NP_001036746.1 | 1 - 2 | 1077 | 64 | |
NM_001275918.1 | NP_001262847.1 | 1 - 2 | 1077 | 64 | ||
NM_170037.2 | NP_732790.3 | 1 - 2 | 1077 | 64 | ||
NM_170038.2 | NP_732791.2 | 1 - 2 | 1077 | 64 | ||
GenBank mRNA | AB190214.1 | - | 1 - 2 | 483 | 64 | |
BT099586.1 | - | 1 - 2 | 1031 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |