Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260322.1 | NP_001247251.1 | 10 - 11 | 2241 | 68 | |
NM_170043.2 | NP_732807.1 | 9 - 10 | 1778 | 68 | ||
GenBank mRNA | AY058312.1 | - | 9 - 10 | 1741 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |