Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir94f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170075.2 | NP_732868.2 | 1 | 1628 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |