Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104422.3 | NP_001097892.1 | 8 - 9 | 5104 | 89 | |
NM_001104423.2 | NP_001097893.1 | 9 - 10 | 4579 | 89 | ||
NM_001275959.1 | NP_001262888.1 | 9 - 10 | 4579 | 89 | ||
NM_176549.3 | NP_788726.2 | 8 - 9 | 4387 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster Inventoried |