Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014647.3 | NP_001014647.2 | 9 - 10 | 2743 | 82 | |
NM_001014648.3 | NP_001014648.2 | 9 - 10 | 2743 | 82 | ||
NM_001014649.3 | NP_001014649.2 | 9 - 10 | 2743 | 82 | ||
NM_001014650.3 | NP_001014650.2 | 9 - 10 | 2743 | 82 | ||
NM_001275976.1 | NP_001262905.1 | 9 - 10 | 2501 | 82 | ||
NM_001275977.2 | NP_001262906.1 | 9 - 10 | 3210 | 82 | ||
NM_001300572.1 | NP_001287501.1 | 9 - 10 | 2743 | 82 | ||
NM_079754.4 | NP_524478.4 | 9 - 10 | 2743 | 82 | ||
NM_170138.3 | NP_732976.1 | 9 - 10 | 2501 | 82 | ||
GenBank mRNA | BT133083.1 | - | 9 - 10 | 2740 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |