Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nAChRalpha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079758.3 | NP_524482.1 | 4 - 5 | 1269 | 72 | |
NM_170146.2 | NP_733001.1 | 5 - 6 | 1201 | 72 | ||
GenBank mRNA | AY058446.1 | - | 4 - 5 | 1255 | 72 | |
X52274.1 | - | 1240 | 72 | |||
X53583.1 | - | 1091 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |