Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nAChRbeta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079759.4 | NP_524483.1 | 7 - 8 | 1135 | 57 | |
NM_170147.3 | NP_733002.1 | 6 - 7 | 3170 | 57 | ||
GenBank mRNA | BT133158.1 | - | 6 - 7 | 3150 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |