Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176558.3 | NP_788735.1 | 2 - 3 | 408 | 58 | |
GenBank mRNA | AY051803.1 | - | 2 - 3 | 361 | 58 | |
U73809.2 | - | 2 - 3 | 361 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |