Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ymp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170230.2 | NP_733109.1 | 3 - 4 | 269 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |