Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170236.2 | NP_733115.1 | 3 - 4 | 886 | 67 | |
GenBank mRNA | BT150256.1 | - | 3 - 4 | 886 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |