Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG33970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038981.2 | NP_001034070.1 | 4 - 5 | 890 | 67 | |
NM_001038982.2 | NP_001034071.1 | 8 - 9 | 2205 | 67 | ||
NM_001300613.1 | NP_001287542.1 | 8 - 9 | 2205 | 67 | ||
GenBank mRNA | BT003531.1 | - | 8 - 9 | 2202 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |