Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lerp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276067.1 | NP_001262996.1 | 2 - 3 | 112 | 59 | |
NM_170288.3 | NP_733167.2 | 2 - 3 | 127 | 59 | ||
GenBank mRNA | BT021416.1 | - | 2 - 3 | 92 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |