Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104420.3 | NP_001097890.1 | 9 - 10 | 1565 | 80 | |
NM_079742.4 | NP_524466.1 | 9 - 10 | 1637 | 80 | ||
GenBank mRNA | AF091624.1 | - | 9 - 10 | 1542 | 80 | |
AY069632.1 | - | 9 - 10 | 1613 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |