Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043286.2 | NP_001036751.1 | 9 - 10 | 4399 | 73 | |
NM_001260355.2 | NP_001247284.1 | 9 - 10 | 4591 | 73 | ||
NM_001275980.1 | NP_001262909.1 | 10 - 11 | 4441 | 73 | ||
NM_079756.3 | NP_524480.2 | 8 - 9 | 4270 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |