Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E(spl)m3-HLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079785.4 | NP_524509.2 | 1 | 950 | 148 | |
GenBank mRNA | AY094858.1 | - | 1 | 945 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |