Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ser | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300629.1 | NP_001287558.1 | 4 - 5 | 1250 | 59 | |
NM_079803.3 | NP_524527.3 | 4 - 5 | 978 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |