Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: side | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144645.3 | NP_001138117.1 | 13 - 14 | 3397 | 63 | |
NM_001260405.2 | NP_001247334.1 | 13 - 14 | 3086 | 63 | ||
NM_001260406.2 | NP_001247335.1 | 13 - 14 | 3819 | 63 | ||
NM_079806.4 | NP_524530.2 | 13 - 14 | 3819 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |