Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets98B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300641.1 | NP_001287570.1 | 2 - 3 | 900 | 81 | |
NM_079811.4 | NP_524535.2 | 2 - 3 | 1376 | 81 | ||
GenBank mRNA | BT053684.1 | - | 2 - 3 | 1348 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |