Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc98D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170288.1 | NP_001163759.1 | 9 - 10 | 3166 | 112 | |
NM_001276107.1 | NP_001263036.1 | 3 - 4 | 848 | 112 | ||
NM_079817.3 | NP_524541.3 | 10 - 11 | 3187 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |