Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkc98E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300647.1 | NP_001287576.1 | 7 - 8 | 1011 | 115 | |
NM_001300648.1 | NP_001287577.1 | 7 - 8 | 1045 | 115 | ||
NM_079821.4 | NP_524545.2 | 7 - 8 | 1011 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |