Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144655.3 | NP_001138127.1 | 2 - 3 | 377 | 72 | |
NM_079843.4 | NP_524582.1 | 2 - 3 | 180 | 72 | ||
NM_170460.2 | NP_733339.1 | 2 - 3 | 349 | 72 | ||
NM_170461.3 | NP_733340.1 | 2 - 3 | 126 | 72 | ||
GenBank mRNA | AY070656.1 | - | 2 - 3 | 126 | 72 | |
BT011442.1 | - | 2 - 3 | 350 | 72 | ||
KX671633.1 | - | 2 - 3 | 130 | 72 | ||
U92431.1 | - | 1 - 2 | 91 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Endogenous Controls Recommended Assay Drosophila melanogaster Inventoried |