Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap-r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079858.4 | NP_524597.1 | 5 - 6 | 2844 | 75 | |
NM_170529.2 | NP_733408.2 | 4 - 5 | 2753 | 75 | ||
GenBank mRNA | AF145647.1 | - | 5 - 6 | 2832 | 75 | |
DQ062780.1 | - | 5 - 6 | 2715 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |