Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276205.1 | NP_001263134.1 | 5 - 6 | 1709 | 64 | |
NM_079866.3 | NP_524605.1 | 5 - 6 | 1730 | 64 | ||
GenBank mRNA | BT126010.1 | - | 5 - 6 | 1508 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |