Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gammaCOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170313.1 | NP_001163784.1 | 4 - 5 | 2441 | 103 | |
NM_001276216.1 | NP_001263145.1 | 4 - 5 | 2441 | 103 | ||
NM_079869.3 | NP_524608.1 | 4 - 5 | 2444 | 103 | ||
NM_170553.2 | NP_733432.1 | 3 - 4 | 2663 | 103 | ||
GenBank mRNA | BT001628.1 | - | 3 - 4 | 2663 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |