Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: disp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079995.3 | NP_524734.2 | 5 - 6 | 2840 | 88 | |
GenBank mRNA | BT050429.1 | - | 5 - 6 | 2840 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |