Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276100.1 | NP_001263029.1 | 4 - 5 | 4246 | 58 | |
NM_170360.2 | NP_733239.1 | 4 - 5 | 4258 | 58 | ||
GenBank mRNA | BT025221.1 | - | 4 - 5 | 4254 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |