Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPsyngamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079826.4 | NP_524550.1 | 2 | 1022 | 127 | |
NM_170425.4 | NP_733304.1 | 2 | 1337 | 127 | ||
NM_170426.3 | NP_733305.1 | 1 | 1097 | 127 | ||
GenBank mRNA | AY113454.1 | - | 2 | 1310 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |