Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144652.2 | NP_001138124.2 | 2 - 3 | 614 | 69 | |
NM_170434.4 | NP_733313.2 | 2 - 3 | 614 | 69 | ||
GenBank mRNA | BT024267.1 | - | 2 - 3 | 564 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |