Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spn-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079844.4 | NP_524583.1 | 2 | 1186 | 132 | |
NM_170463.3 | NP_733342.1 | 1 | 1249 | 132 | ||
GenBank mRNA | BT089037.1 | - | 2 | 1182 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |