Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PH4alphaSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170497.4 | NP_733376.1 | 4 - 5 | 1385 | 84 | |
GenBank mRNA | BT023117.1 | - | 4 - 5 | 1358 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |