Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PH4alphaPV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170516.1 | NP_733395.1 | 7 - 8 | 1568 | 76 | |
GenBank mRNA | AF495539.1 | - | 7 - 8 | 1568 | 76 | |
BT150220.1 | - | 7 - 8 | 1568 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |