Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stops | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170309.2 | NP_001163780.1 | 3 - 4 | 863 | 87 | |
NM_170537.2 | NP_733416.2 | 2 - 3 | 679 | 87 | ||
NM_170538.3 | NP_733417.1 | 3 - 4 | 818 | 87 | ||
GenBank mRNA | AY047558.1 | - | 2 - 3 | 435 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |