Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275391.1 | NP_001262320.1 | 6 - 7 | 1924 | 78 | |
NM_176409.2 | NP_788586.1 | 6 - 7 | 1924 | 78 | ||
GenBank mRNA | AY121627.1 | - | 6 - 7 | 1924 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |