Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275975.1 | NP_001262904.1 | 2 - 3 | 1410 | 76 | |
NM_176556.2 | NP_788733.1 | 2 - 3 | 1410 | 76 | ||
NM_176557.2 | NP_788734.1 | 2 - 3 | 1410 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |