Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260351.1 | NP_001247280.1 | 9 - 10 | 4348 | 82 | |
NM_001275974.1 | NP_001262903.1 | 8 - 9 | 3145 | 82 | ||
NM_001275975.1 | NP_001262904.1 | 8 - 9 | 4312 | 82 | ||
NM_001300571.1 | NP_001287500.1 | 7 - 8 | 3106 | 82 | ||
NM_176556.2 | NP_788733.1 | 8 - 9 | 4312 | 82 | ||
NM_176557.2 | NP_788734.1 | 8 - 9 | 4312 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |