Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170289.2 | NP_001163760.1 | 5 - 6 | 1975 | 73 | |
NM_001170290.2 | NP_001163761.1 | 5 - 6 | 1975 | 73 | ||
NM_176574.3 | NP_788751.2 | 5 - 6 | 1975 | 73 | ||
NM_176575.3 | NP_788752.2 | 5 - 6 | 1975 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |