Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap-H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206471.2 | NP_996193.1 | 8 - 9 | 2543 | 66 | |
GenBank mRNA | BT001334.1 | - | 8 - 9 | 2543 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |