Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo95E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032033.2 | NP_001027204.2 | 5 - 6 | 1469 | 133 | |
NM_206555.2 | NP_996278.1 | 4 - 5 | 1572 | 133 | ||
NM_206556.2 | NP_996279.1 | 4 - 5 | 1572 | 133 | ||
GenBank mRNA | AF454350.1 | - | 4 - 5 | 1572 | 133 | |
AF454351.1 | - | 4 - 5 | 1572 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |