Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo95E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032033.2 | NP_001027204.2 | 7 - 8 | 1640 | 66 | |
NM_001032035.2 | NP_001027206.1 | 6 - 7 | 1759 | 66 | ||
NM_001032036.2 | NP_001027207.1 | 7 - 8 | 1656 | 66 | ||
NM_001032037.2 | NP_001027208.1 | 7 - 8 | 1773 | 66 | ||
NM_206555.2 | NP_996278.1 | 6 - 7 | 1743 | 66 | ||
NM_206556.2 | NP_996279.1 | 6 - 7 | 1743 | 66 | ||
GenBank mRNA | AF454350.1 | - | 6 - 7 | 1743 | 66 | |
AF454351.1 | - | 6 - 7 | 1743 | 66 | ||
BT011452.1 | - | 7 - 8 | 1653 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |