Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170162.2 | NP_001163633.1 | 2 - 3 | 246 | 86 | |
NM_206501.3 | NP_996223.1 | 2 - 3 | 573 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |