Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LysB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274316.1 | NP_001261245.1 | 1 | 429 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |