Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsc70-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079339.3 | NP_524063.1 | 1 - 2 | 324 | 146 | |
GenBank mRNA | L01501.1 | - | 1 - 2 | 462 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |