Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079562.3 | NP_524286.1 | 2 - 3 | 641 | 84 | |
NM_169261.3 | NP_731319.1 | 3 - 4 | 504 | 84 | ||
NM_169262.2 | NP_731320.1 | 2 - 3 | 718 | 84 | ||
GenBank mRNA | BT001362.1 | - | 2 - 3 | 718 | 84 | |
BT001636.1 | - | 3 - 4 | 460 | 84 | ||
J04725.1 | - | 2 - 3 | 638 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |