Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132013.3 | NP_572241.2 | 1 - 2 | 243 | 146 | |
GenBank mRNA | BT011040.1 | - | 2114 | 146 | ||
BT099555.1 | - | 1 - 2 | 252 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |