Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp59a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176249.3 | NP_788429.1 | 1 - 2 | 385 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |