Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ama | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057231.4 | NP_476579.1 | 2 | 1482 | 108 | |
NM_169160.1 | NP_731114.2 | 2 | 1773 | 108 | ||
NM_176412.2 | NP_788589.1 | 2 | 1466 | 108 | ||
GenBank mRNA | AY051911.1 | - | 2 | 1470 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |