Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ama | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057231.4 | NP_476579.1 | 2 | 540 | 67 | |
NM_169160.1 | NP_731114.2 | 2 | 831 | 67 | ||
NM_176412.2 | NP_788589.1 | 2 | 524 | 67 | ||
GenBank mRNA | AY051911.1 | - | 2 | 528 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |