Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144545.2 | NP_001138017.1 | 1 | 744 | 71 | |
NM_057446.3 | NP_476794.1 | 2 | 739 | 71 | ||
GenBank mRNA | BT028810.2 | - | 1 | 751 | 71 | |
BT028840.2 | - | 1 | 751 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |