Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273374.2 | NP_001260303.1 | 2 | 1083 | 88 | |
NM_001298856.1 | NP_001285785.1 | 2 | 853 | 88 | ||
NM_057526.5 | NP_476874.1 | 2 | 822 | 88 | ||
GenBank mRNA | AY094799.1 | - | 2 | 802 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |