Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlp84B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316489.1 | NP_001303418.1 | 2 | 1047 | 60 | |
NM_057774.5 | NP_477122.1 | 2 | 1047 | 60 | ||
NM_169173.3 | NP_731134.1 | 2 | 1030 | 60 | ||
GenBank mRNA | BT003202.1 | - | 2 | 1044 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |